missing translation for 'onlineSavingsMsg'
Learn More

Myoglobin Antibody (3F7), Novus Biologicals™

Product Code. 18341588 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341588 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341588 Supplier Novus Biologicals Supplier No. H00004151M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myoglobin Monoclonal antibody specifically detects Myoglobin in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myoglobin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3F7
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Alias MGC13548, myoglobin, PVALB
Host Species Mouse
Immunogen Native purified human MB.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 4151
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.