missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myogenin Mouse anti-Human, Porcine, Rat, Clone: 1C10B9, ProteinTech
Mouse Monoclonal Antibody
Brand: Proteintech 67082-1-IG-150UL
This item is not returnable.
View return policy
Description
Aliquoting is unnecessary for -20°C storage.
Myogenin Monoclonal antibody specifically detects Myogenin in Human, Porcine, Rat samples. It is validated for Western BlotSpecifications
| Myogenin | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P15173, P20428, P49812 | |
| MYOG | |
| Myogenin Fusion Protein Ag6414 | |
| 150 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1C10B9 | |
| Unconjugated | |
| MYOG | |
| bHLHc3; Class C basic helix-loop-helix protein 3; MYF4; Myf-4; myo; MYOD1-related protein; MYOG; Myogenic factor 4; Myogenic factor-4; myogenin; myogenin (myogenic factor 4) | |
| Mouse | |
| Protein G | |
| RUO | |
| 29148, 4656, 497618 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction