missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myogenin Monoclonal Antibody (F5D)

Description
MA5-11486 targets Myogenin in GS, IF, IHC (P), IP, and WB applications and shows reactivity with Feline, Human, mouse, and Rat samples. The MA5-11486 immunogen is recombinant protein containing rat myogenin aa 30-224.
Myogenin is a muscle-specific transcription factor that can induce myogenesis in a variety of cell types in tissue culture. It is a member of a large family of proteins related by sequence homology, the helix-loop-helix proteins. It is essential for the development of functional skeletal muscle.
Specifications
Specifications
| Antigen | Myogenin |
| Applications | Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | F5D |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | MYOG |
| Gene Accession No. | P12979, P15173, P20428 |
| Gene Alias | bHLHc3; Class C basic helix-loop-helix protein 3; MYF4; Myf-4; myo; MYOD1-related protein; MYOG; Myogenic factor 4; Myogenic factor-4; myogenin; myogenin (myogenic factor 4) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?