missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myogenin Antibody (MGN185 + F5D) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Myogenin Monoclonal antibody specifically detects Myogenin in Human, Mouse, Rat, Porcine, Feline samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Myogenin |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MGN185 + F5D |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 10 mM PBS |
| Gene Alias | BHLHC3, bHLHc3Myogenic factor-4, myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) |
| Host Species | Mouse |
| Immunogen | Human Myogenin recombinant protein (MGN185); Rat Myogenin recombinant fragment containing amino acid 30-224 (F5D) (Uniprot: P15173) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?