Learn More
Epredia™ Myogenin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Deliver accurate, reproducible results with the Epredia™ Myogenin Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1113-PABX
Code : LD
Additional Details : Weight : 1.00000kg
Description
Myogenin is a member of a family of myogenic regulatory genes, which includes MyoD, myf5 and MRF4. These genes encode a set of transcription factors, which are essential for muscle development. Transfection of myogenin into multipotential mesodermal cells have been shown to convert the mesodermal cells to myoblasts. Expression of myogenin is restricted to cells of skeletal muscle origin. It is therefore a useful marker for tumors of the muscle lineage, being strongly expressed in alveolar rhabdomyosarcomas.Host Species: Mouse
Clone: F5D
Isotype: IgG1/κ
Species Reactivity: Human, Mouse, Rat and Cat. Does not react with quail. Others not known.
Epitope: aa 138-158
Immunogen: Recombinant protein containing rat myogenin aa 30-224
Molecular Weight: 34kDa
Positive Control: Rh30 cells, Rhabdomyosarcoma or Wilm's tumors
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native verified)
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
Myogenin Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Muscle Biology | |
Feline, Human, Mouse, Rat | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
F5D | |
Unconjugated | |
Recombinant protein containing rat myogenin aa 30-224 | |
RUO | |
Primary | |
Concentrated |