missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myofibroblast Marker Mouse anti-Human, Clone: PR 2D3, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Myofibroblast Marker |
|---|---|
| Clone | PR 2D3 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1-2ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Myofibroblast Marker Monoclonal specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Myofibroblast Marker | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Crude homogenate of normal human colorectal mucosa | |
| Primary | |
| Store at 4°C. |
| PR 2D3 | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1-2ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title