missing translation for 'onlineSavingsMsg'
Learn More

Myofibroblast Marker Mouse anti-Human, Clone: PR 2D3, Novus Biologicals™

Product Code. 18358132 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358132 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358132 Supplier Novus Biologicals Supplier No. NBP30774520UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myofibroblast Marker Monoclonal specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myofibroblast Marker
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PR 2D3
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml
Formulation 10 mM PBS with 0.05% BSA
Host Species Mouse
Immunogen Crude homogenate of normal human colorectal mucosa
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.