missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOD1 Mouse anti-Human, Clone: 5D1D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66214-1-IG-150UL
This item is not returnable.
View return policy
Description
The Myogenic determination gene (MyoD) was first identified by the virtue of its ability to convert embryonic mouse fibroblast cells to muscle cells. It was subsequently shown that forced expression of MyoD (human homolog is myf 3) gene in a wide variety of normal and neoplastic cells could either convert the cells to muscle cells or activate a set of the otherwise transcriptionally inactive muscle-specific genes in these cells. The regulatory domain of the MyoD gene product lies within a 70 amino acid region and comprises a basic DNA binding motif and a helix-loop-helix (HLH) dimerization motif. Subsequent studies identified three other genes whose products shared sequence homology for the basic HLH domain of MyoD. These are; myf5, myogenin (human homolog is myf4) and myf6 (also known as MRF4 and herculin).Specifications
| MYOD1 | |
| Monoclonal | |
| 1.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15172 | |
| MYOD1 | |
| MYOD1 Fusion Protein Ag13512 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 5D1D12 | |
| Unconjugated | |
| MYOD1 | |
| bHLHc1, Myf 3, MYF3, MYOD, MYOD1, myogenic differentiation 1, Myogenic factor 3, PUM | |
| Mouse | |
| Protein G | |
| RUO | |
| 4654 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction