missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyoD Mouse anti-Human, Clone: rMYD712, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00
Spezifikation
| Antigen | MyoD |
|---|---|
| Clone | rMYD712 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Beschreibung
MyoD Monoclonal specifically detects MyoD in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Spezifikation
| MyoD | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 4654 | |
| IgG1 κ | |
| Protein A or G purified |
| rMYD712 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
| Recombinant full-length human MyoD protein | |
| Primary | |
| Store at 4°C. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts