missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyoD Mouse anti-Human, Clone: rMYD712, Novus Biologicals™
Mouse Monoclonal Antibody
£387.00
Specifications
| Antigen | MyoD |
|---|---|
| Clone | rMYD712 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | ELISA, Peptide Array |
Description
MyoD Monoclonal specifically detects MyoD in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| MyoD | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 4654 | |
| IgG1 κ | |
| Protein A or G purified |
| rMYD712 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
| Recombinant full-length human MyoD protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title