missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MyoD Monoclonal specifically detects MyoD in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | MyoD |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MYOD1/3418R |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin : 1-2 ug/mL |
| Formulation | 10mM PBS |
| Gene Alias | bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?