missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MyoD Antibody (MYOD1/2075R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-75772CL1
This item is not returnable.
View return policy
Description
MyoD Monoclonal antibody specifically detects MyoD in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Protein Array
Specifications
| MyoD | |
| Monoclonal | |
| CoraFluor 1 | |
| bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
| Recombinant human full-length MyoD protein (Uniprot: P15172 ) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific | |
| 4654 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| MYOD1/2075R | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction