missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYOC Polyclonal Antibody
GREENER_CHOICE

Product Code. 16678904
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16678904 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16678904 Supplier Invitrogen™ Supplier No. PA5102433

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYOC.

Myocilin is an extracellular protein expressed in the eye, including the retina, trabecular meshwork and ciliary body. Myocilin can form homomultimers in vivo and can also associate with components of the ECM via interactions with the Hep II domain of FibroPVRL1. In addition, myocilin interacts with myosin regulatory light chain, a component of the myosin motor protein complex. This interaction implies a role for myocilin in the actomyosin system, linking myocilin to the functional status of the trabecular meshwork (TM), which is responsible for controlling the intraocular pressure (IOP). Alterations in functions of the TM may lead to IOP elevation and development of glaucoma, a major cause of blindness. Myocilin is encoded by MYOC (also designated TIGR), a gene that maps to the GLC1A locus on chromosome 1q24. 3 and is susceptible to mutations. Mutations in the MYOC gene are specifically linked with primary open angle glaucoma (POAG), a blinding disease characterized by progressive loss of retinal ganglion cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYOC
Gene Accession No. O70624, Q99972, Q9R1J4
Gene Alias AI957332; GLC1A; GPOA; JOAG; JOAG1; juvenile-onset open-angle glaucoma 1; mutated trabecular meshwork-induced glucocorticoid response protein; MYOC; Myocilin; Myocilin 20 kDa N-terminal fragment; Myocilin 35 kDa N-terminal fragment; Myocilin 55 kDa subunit; myocilin trabecular meshwork inducible glucocorticoid response; myocilin trabecular meshwork inducible glucocorticoid response protein; Myocilin, C-terminal fragment; Myocilin, N-terminal fragment; myocilin, trabecular meshwork inducible glucocorticoid response; TIGR; trabecular meshwork inducible glucocorticoid response protein; trabecular meshwork-induced glucocorticoid response protein
Gene Symbols MYOC
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYOC(Accession Q99972), corresponding to amino acid residues R342-Y392.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17926, 4653, 81523
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.