missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYOC Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA537381

Product Code. 15564371

  • £472.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects endogenous protein at a molecular weight of 57 kDa. Purity is >95% by SDS-PAGE.

Myocilin is an extracellular protein expressed in the eye, including the retina, trabecular meshwork and ciliary body. Myocilin can form homomultimers in vivo and can also associate with components of the ECM via interactions with the Hep II domain of FibroPVRL1. In addition, myocilin interacts with myosin regulatory light chain, a component of the myosin motor protein complex. This interaction implies a role for myocilin in the actomyosin system, linking myocilin to the functional status of the trabecular meshwork (TM), which is responsible for controlling the intraocular pressure (IOP). Alterations in functions of the TM may lead to IOP elevation and development of glaucoma, a major cause of blindness. Myocilin is encoded by MYOC (also designated TIGR), a gene that maps to the GLC1A locus on chromosome 1q24. 3 and is susceptible to mutations. Mutations in the MYOC gene are specifically linked with primary open angle glaucoma (POAG), a blinding disease characterized by progressive loss of retinal ganglion cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MYOC
Polyclonal
Unconjugated
MYOC
AI957332; GLC1A; GPOA; JOAG; JOAG1; juvenile-onset open-angle glaucoma 1; mutated trabecular meshwork-induced glucocorticoid response protein; MYOC; Myocilin; Myocilin 20 kDa N-terminal fragment; Myocilin 35 kDa N-terminal fragment; Myocilin 55 kDa subunit; myocilin trabecular meshwork inducible glucocorticoid response; myocilin trabecular meshwork inducible glucocorticoid response protein; Myocilin, C-terminal fragment; Myocilin, N-terminal fragment; myocilin, trabecular meshwork inducible glucocorticoid response; TIGR; trabecular meshwork inducible glucocorticoid response protein; trabecular meshwork-induced glucocorticoid response protein
Rabbit
Antigen Affinity Chromatography
RUO
17926, 4653
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O70624, Q99972
MYOC
Recombinant full length human MYOC.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.