Learn More
Invitrogen™ MYO6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110004
Description
Immunogen sequence: IAQSEAELIS DEAQADLALR RNDGTRPKMT PEQMAKEMSE FLSRGPAVLA TKAAAGTKKY DLSKWKYAEL RDTINTSCDI ELLAACREEF HRRLKVYHAW KSKNKKRNTE TEQRAPKSVT DYDFAPFLNN SPQQNPAAQI PARQREIEMN RQQRFFRIPF IRPADQYKDP QSKKKGWWYA HFDGPWIARQ MELHPDKPPI LLVAGKDDME MCELNLEETG LTRKRGAEIL PRQFEEIWER CGGIQYLQNA IESRQARPTY ATAMLQSLLK.
This gene encodes a protein involved intracellular vesicle and organelle transport, especially in the hair cell of the inner ear. Mutations in this gene have been found in patients with non-syndromic autosomal dominant and recessive hearing loss.
Specifications
| MYO6 | |
| Polyclonal | |
| Unconjugated | |
| MYO6 | |
| BC029719; DFNA22; DFNB37; KIAA0389; MYO6; myosin; myosin VI; myosin-VI; RGD1560646; Snell's waltzer; sv; tailchaser; Tlc; unconventional myosin; unconventional myosin-6; Unconventional myosin-VI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17920, 315840, 4646 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.24 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q64331, Q9UM54 | |
| MYO6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1016-1285 of human MYO6 (NP_004990.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.