missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYO6 Monoclonal Antibody (K70041_12A6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556268

 View more versions of this product

Product Code. 30212649

  • £760.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: EEAERLRRIQ EEMEKERKRR EEDEKRRRKE EEERRMKLEM EAKRKQEEEE RKKREDDEKR IQAEVEAQLA RQKEEESQQQ AVLEQERRDR ELALRIAQSE AELISDEAQA DLALRRSLDS YPVSKNDGTR PKMTPEQMAK EMSEFLSRGP AVLATKAAAG TKKYDLSKWK YAELRDTINT SCDIELLAAC REEFHRRLKV YHAWKSKNKK RNTETEQRAP KSVTDYDFAP FLNNSPQQNP AAQIPARQRE IEMNRQQRFF RIPFIRPADQ YKDPQSKKKG WWYAHFDGPW IARQMELHPD KPPILLVAGK DDMEMCELNL EETGLTRKRG AEILPRQFEE

This gene encodes a protein involved intracellular vesicle and organelle transport, especially in the hair cell of the inner ear. Mutations in this gene have been found in patients with non-syndromic autosomal dominant and recessive hearing loss.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MYO6
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q9UM54
MYO6
Recombinant human MYO6 (immunogen range is 921-1260).
200 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
K70041_12A6
Unconjugated
MYO6
BC029719; DFNA22; DFNB37; KIAA0389; MYO6; myosin; myosin VI; myosin-VI; RGD1560646; Snell's waltzer; sv; tailchaser; Tlc; unconventional myosin; unconventional myosin-6; Unconventional myosin-VI
Mouse
Protein A
RUO
4646
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.