missing translation for 'onlineSavingsMsg'
Learn More

MYO6 Antibody (2E12), Novus Biologicals™

Product Code. 18340008 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340008 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340008 Supplier Novus Biologicals Supplier No. H00004646M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MYO6 Monoclonal antibody specifically detects MYO6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO6
Applications Western Blot, ELISA, Immunohistochemistry, Sandwich ELISA
Classification Monoclonal
Clone 2E12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004990.2
Gene Alias DFNA22, DFNB37, KIAA0389deafness, autosomal recessive 37, myosin VI, myosin-VI, Unconventional myosin-6
Host Species Mouse
Immunogen MYO6 (NP_004990.2, 1188 a.a. ~ 1285 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KKKGWWYAHFDGPWIARQMELHPDKPPILLVAGKDDMEMCELNLEETGLTRKRGAEILPRQFEEIWERCGGIQYLQNAIESRQARPTYATAMLQSLLK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4646
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.