missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MYO5C 293T Cell Transient Overexpression Lysate

Product Code. 18378809 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
This item is not returnable. View return policy

Product Code. 18378809

Brand: Novus Biologicals™ H00055930T02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Research Category Cell Biology, Cytoskeleton Markers, Endocrinology, Signal Transduction
Target MYO5C
Synonym MGC74969, myosin 5C, myosin VC, myosin-Vc
Gene Symbol MYO5C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.