missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110003
This item is not returnable.
View return policy
Description
Immunogen sequence: LTNLEGIYNS ETEKLRSDLE RLQLSEEEAK VATGRVLSLQ EEIAKLRKDL EQTRSEKKCI EEHADRYKQE TEQLVSNLKE ENTLLKQEKE ALNHRIVQQA KEMTETMEKK LVEETKQLEL DLNDERLRYQ NLLNEFSRLE ERYDDLKEEM TLMVHVPKPG HKRTDSTHSS NESEYIFSSE IAEMEDIPSR TEEPSEKKVP L.
This gene is one of three myosin V heavy-chain genes, belonging to the myosin gene superfamily. Myosin V is a class of actin-based motor proteins involved in cytoplasmic vesicle transport and anchorage, spindle-pole alignment and mRNA translocation. The protein encoded by this gene is abundant in melanocytes and nerve cells. Mutations in this gene cause Griscelli syndrome type-1 (GS1), Griscelli syndrome type-3 (GS3) and neuroectodermal melanolysosomal disease, or Elejalde disease. Multiple alternatively spliced transcript variants encoding different isoforms have been reported, but the full-length nature of some variants has not been determined.
Specifications
| MYO5A | |
| Polyclonal | |
| Unconjugated | |
| MYO5A | |
| 9630007J19Rik; AI413174; AI661011; D; d-120J; Dbv; Dilute; dilute lethal-20J protein; dilute myosin heavy chain, non-muscle; dilute-opisthotonus; Dop; flail; flailer; flr; GS1; MVa; MYH12; MYO5; Myo5a; myosin 5a; Myosin heavy chain 12; Myosin heavy chain p190; myosin I heavy chain isoform; myosin V; myosin VA; myosin VA (heavy chain 12, myosin); myosin VA (heavy chain 12, myoxin); myosin VA (heavy polypeptide 12, myoxin); myosin, heavy polypeptide kinase; myosin-12; Myosin-V; myosin-Va; MyoVA; myoxin; MYR12; OTTMUSP00000017756; OTTMUSP00000019176; OTTMUSP00000046608; p190 myosin heavy chain; Sev-1; unconventional myosin-Va | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17918, 25017, 4644 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.86 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99104, Q9QYF3, Q9Y4I1 | |
| MYO5A | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction