Learn More
Invitrogen™ MYO3B (MYO3 beta) Recombinant Human Protein
MYO3B (MYO3 beta) is a member of the Class III myosins that are actin-dependent motor proteins containing an amino-terminal kinase domain. Recombinant human MYO3B (1-326) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Brand: Invitrogen™ PV6368
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MYO3B contains a N-terminal kinase domain, followed by motor, neck, and tail domains. The MYO3B gene generates a variety of splice variants that contain 1 or 2 calmodulin-binding (IQ) motifs in the neck domain and 1 of 3 domains in the tail domain. Northern blot analysis shows expression of a 7-kb MYO3B transcript in the human retina but not in a RPE cell line. The MYO3B gene transcript is also detected in the kidney, intestine and testis. The MYO3B gene maps to chromosome 2q31.1-q31.2 by genomic sequence alignment.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
63.3kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
aa 1-362 | |
Recombinant |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol | |
100μg | |
myosin IIIB | |
Human | |
GST Tag |