missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO1C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544537
This item is not returnable.
View return policy
Description
Peptide sequence: NPVLEAFGNA KTLRNDNSSR FGKYMDVQFD FKGAPVGGHI LSYLLEKSRV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
MYO1C is a member of the unconventional myosin protein family, which are actin-based molecular motors. The protein is found in the cytoplasm, and one isoform with a unique N-terminus is also found in the nucleus. The nuclear isoform associates with RNA polymerase I and II and functions in transcription initiation. The mouse ortholog of this protein also functions in intracellular vesicle transport to the plasma membrane.
Specifications
| MYO1C | |
| Polyclonal | |
| Unconjugated | |
| MYO1C | |
| C80397; FLJ23903; mm1beta; MMIb; MMI-beta; myo1c; myo1ca; MYO1E; myosin 1c, paralog a; myosin heavy chain myr 2; myosin I; Myosin I beta; myosin IB; myosin IC; myosin Ic, paralog a; myosin IC-like; myosin-I beta; myosin-Ibeta; myosin-Ic; Myr2; NMI; nuclear myosin I; nuclear myosin I beta; si:ch211-69i14.4; unconventional myosin Myr2 I heavy chain; unconventional myosin-Ic | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4641 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00159 | |
| MYO1C | |
| Synthetic peptide directed towards the N-terminal of human MYO1C (aa 178-227). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction