missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO1B Polyclonal Antibody

MYO1B Polyclonal Antibody
Brand: Invitrogen PA5142637
This item is not returnable.
View return policy
Description
MYO1B Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| MYO1B | |
| Polyclonal | |
| Unconjugated | |
| MYO1B | |
| AA406997; MIH-L; MMIa; MMI-alpha; molecular motor; MYH-1c; Myo1a; MYO1B; MYO1B variant protein; myosin heavy chain myr 1; Myosin I alpha; Myosin IA (Myosin I alpha) (MMI-alpha) (MMIa) (MIH-L); myosin Ib; myosin-I alpha; myosin-Ib; myr1; Unconventional myosin-Ib | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 17912, 4430 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O43795, P46735 | |
| MYO1B | |
| Peptide with sequence C-RTVEAKQEKVSTT. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction