missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO18B Monoclonal Antibody (1683CT270.39.50)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524752
This item is not returnable.
View return policy
Description
MYO18B Monoclonal Antibody for Western Blot, IHC (P), Flow
The protein encoded by this gene may regulate muscle-specific genes when in the nucleus and may influence intracellular trafficking when in the cytoplasm. The encoded protein functions as a homodimer and may interact with F actin. Mutations in this gene are associated with lung cancer.
Specifications
| MYO18B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8IUG5 | |
| MYO18B | |
| human MYO18B recombinant protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1683CT270.39.50 | |
| Unconjugated | |
| MYO18B | |
| DKFZp434C2422; DKFZp779C1668; FLJ41942; KFS4; MY18B; MYO18B; myosin 18B; myosin XVIIIB; Myosin-XVIIIb; unconventional myosin-XVIIIb | |
| Mouse | |
| Protein G | |
| RUO | |
| 84700 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction