missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO18A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA554328
This item is not returnable.
View return policy
Description
Immunogen sequence: RFSFSQRSRD ESASETSTPS EHSAAPSPQV EVRTLEGQLV QHPGPGIPRP GHRSRAPELV TKKFPVDLRL PPVVPLPPPT LRELELQRRP TGDFGFSLRR TTMLDRGPEG QACRRVVHFA EP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
MYO18A may link Golgi membranes to the cytoskeleton and participate in the tensile force required for vesicle budding from the Golgi. Thereby, may play a role in Golgi membrane trafficking and could indirectly give its flattened shape to the Golgi apparatus. Alternatively, in concert with LURAP1 and CDC42BPA/CDC42BPB, has been involved in modulating lamellar actomyosin retrograde flow that is crucial to cell protrusion and migration. May be involved in the maintenance of the stromal cell architectures required for cell to cell contact. Regulates trafficking, expression, and activation of innate immune receptors on macrophages. Plays a role to suppress inflammatory responsiveness of macrophages via a mechanism that modulates CD14 trafficking.
Specifications
| MYO18A | |
| Polyclonal | |
| Unconjugated | |
| MYO18A | |
| CD245; KIAA0216; MAJN; molecule associated with JAK3 N-terminus; Myo18a; MyoPDZ; myosin 18a; myosin containing a PDZ domain; myosin containing PDZ domain; myosin XVIIIa; myosin XVIIIb; myosin-XVIIIa; MysPDZ; SP-A receptor subunit SP-R210 alphaS; SPR210; SP-R210; Surfactant protein receptor SP-R210; Unconventional myosin-XVIIIa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 399687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92614 | |
| MYO18A | |
| Recombinant protein corresponding to Human MYO18A | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction