missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO18A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576549
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
MYO18A may link Golgi membranes to the cytoskeleton and participate in the tensile force required for vesicle budding from the Golgi. Thereby, may play a role in Golgi membrane trafficking and could indirectly give its flattened shape to the Golgi apparatus. Alternatively, in concert with LURAP1 and CDC42BPA/CDC42BPB, has been involved in modulating lamellar actomyosin retrograde flow that is crucial to cell protrusion and migration. May be involved in the maintenance of the stromal cell architectures required for cell to cell contact. Regulates trafficking, expression, and activation of innate immune receptors on macrophages. Plays a role to suppress inflammatory responsiveness of macrophages via a mechanism that modulates CD14 trafficking.
Specifications
| MYO18A | |
| Polyclonal | |
| Unconjugated | |
| MYO18A | |
| CD245; KIAA0216; MAJN; molecule associated with JAK3 N-terminus; Myo18a; MyoPDZ; myosin 18a; myosin containing a PDZ domain; myosin containing PDZ domain; myosin XVIIIa; myosin XVIIIb; myosin-XVIIIa; MysPDZ; SP-A receptor subunit SP-R210 alphaS; SPR210; SP-R210; Surfactant protein receptor SP-R210; Unconventional myosin-XVIIIa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 360013, 360570, 399687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92614, Q9JMH9 | |
| MYO18A | |
| Recombinant fusion protein of human MYO18A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction