missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO18A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£296.00 - £423.00
Specifications
| Antigen | MYO18A |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MYO18A Polyclonal specifically detects MYO18A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MYO18A | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| KIAA0216DKFZp686L0243, MAJN, Molecule associated with JAK3 N-terminus, myosin 18A, Myosin containing a PDZ domain, myosin containing PDZ domain, myosin XVIIIA, myosin-XVIIIa, MYSPDZ, SP-A receptor subunit SP-R210 alphaS, SPR210 | |
| MYO18A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q92614 | |
| 399687 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RFSFSQRSRDESASETSTPSEHSAAPSPQVEVRTLEGQLVQHPGPGIPRPGHRSRAPELVTKKFPVDLRLPPVVPLPPPTLRELELQRRPTGDFGFSLRRTTMLDRGPEGQACRRVVHFAEP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title