missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO18A Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79840G
This item is not returnable.
View return policy
Description
MYO18A Polyclonal antibody specifically detects MYO18A in Human samples. It is validated for Western Blot
Specifications
| MYO18A | |
| Polyclonal | |
| Western Blot | |
| KIAA0216DKFZp686L0243, MAJN, Molecule associated with JAK3 N-terminus, myosin 18A, Myosin containing a PDZ domain, myosin containing PDZ domain, myosin XVIIIA, myosin-XVIIIa, MYSPDZ, SP-A receptor subunit SP-R210 alphaS, SPR210 | |
| The immunogen recognized by this antibody maps to a region between residue 2004 andd 2054 of human myosin 18A using the numbering given in entry NP_510880.2 (GeneID 399687). | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 399687 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction