missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567217
This item is not returnable.
View return policy
Description
Immunogen sequence: LFQSKLSQTGS LVSAYPSFKF RGHKSALLSK KMTASSIIGE NKNYLELSKL LKKKGTSTFL QRLERGDPVT IASQLRKSLM Highest antigen sequence identity to the following orthologs - mouse 83%, rat 80%.
Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments. May be involved in targeting of the catalytic subunit of protein phosphatase 1 during brain development. Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
Specifications
| MYO16 | |
| Polyclonal | |
| Unconjugated | |
| Myo16 | |
| BM140241; C230040D10Rik; Gm1105; Kiaa0865; LOW QUALITY PROTEIN: unconventional myosin-XVI; mKIAA0865; MYAP3; MYO16; myo16 {ECO:0000250; MYO16 variant protein; Myo16b; Myosin heavy chain myr 8; myosin XVI; myosin-XVI; myosin-XVI; unconventional myosin-XVI; Myr8; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 3; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 3; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 3; NYAP3; PPP1R107; protein phosphatase 1, regulatory subunit 107; unconventional myosin-16; Unconventional myosin-XVI; UniProtKB:Q9Y6X6} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23026 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6X6 | |
| Myo16 | |
| Recombinant Human MYO16. Recombinant protein control fragment (Product #RP-107852). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction