missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYO16 Polyclonal antibody specifically detects MYO16 in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | MYO16 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | KIAA0865, MYO16 variant protein, MYO16B, myosin XVI, myosin-XVI, MYR8, unconventional myosin-16 |
| Host Species | Rabbit |
| Immunogen | myosin XVI. (Uniprot# Q9Y6X6) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?