missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO15A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144364
This item is not returnable.
View return policy
Description
MYO15A Polyclonal Antibody for ICC/IF, ELISA
This gene encodes an unconventional myosin. This protein differs from other myosins in that it has a long N-terminal extension preceding the conserved motor domain. Studies in mice suggest that this protein is necessary for actin organization in the hair cells of the cochlea. Mutations in this gene have been associated with profound, congenital, neurosensory, nonsyndromal deafness. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Read-through transcripts containing an upstream gene and this gene have been identified, but they are not thought to encode a fusion protein. Several alternatively spliced transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq, Jul 2008].
Specifications
| MYO15A | |
| Polyclonal | |
| Unconjugated | |
| MYO15A | |
| DFNB3; MYO15; MYO15A; myosin XVA; myosin-XV; unconventional myosin-15; Unconventional myosin-XV | |
| Rabbit | |
| Protein G | |
| RUO | |
| 51168 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 3.27 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q9UKN7 | |
| MYO15A | |
| Recombinant Human Unconventional myosin-XV protein (237-451AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction