Learn More
Invitrogen™ MYO10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120164
Description
Positive test controls include: U-87MG, Mouse kidney, Mouse lung. The target is usually found in the following locations: Cell projection, Cytoplasm, Peripheral membrane protein, cell cortex, cytoskeleton, cytosol, filopodium membrane, filopodium tip, lamellipodium, ruffle. Immunogen sequence: EAELRAQQEE ETRKQQELEA LQKSQKEAEL TRELEKQKEN KQVEEILRLE KEIEDLQRMK EQQELSLTEA SLQKLQERRD QELRRLEEEA CRAAQEFLES.
This gene encodes a member of the myosin superfamily. The protein represents an unconventional myosin; it should not be confused with the conventional non-muscle myosin-10 (MYH10). Unconventional myosins contain the basic domains of conventional myosins and are further distinguished from class members by their tail domains. This gene functions as an actin-based molecular motor and plays a role in integration of F-actin and microtubule cytoskeletons during meiosis.
Specifications
| MYO10 | |
| Polyclonal | |
| Unconjugated | |
| MYO10 | |
| AW048724; D15Ertd600e; KIAA0799; mKIAA0799; MYO10; myosin X; myosin-X; myosin-X; unconventional myosin-X; unconventional myosin-10; Unconventional myosin-X; unconventionnal myosin-X | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17909, 4651 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F8VQB6, Q9HD67 | |
| MYO10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 845-944 of human MYO10 (NP_036466.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.