missing translation for 'onlineSavingsMsg'
Learn More

myo-Inositol Hexaacetate, TRC

Product Code. 30397765
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397765 1 g -
30392177 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397765 Supplier Toronto Research Chemicals Supplier No. TRCI6660001G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1254-38-2
Molecular Formula C18 H24 O12
Molecular Weight (g/mol) 432.38
IUPAC Name (2,3,4,5,6-pentaacetyloxycyclohexyl) acetate
SMILES CC(=O)O[C@@H]1[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H24O12/c1-7(19)25-13-14(26-8(2)20)16(28-10(4)22)18(30-12(6)24)17(29-11(5)23)15(13)27-9(3)21/h13-18H,1-6H3/t13-,14-,15-,16+,17-,18-
Formula Weight 432.1268
Chemical Name or Material myo-Inositol Hexaacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.