missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MYLK4, GST Tag Recombinant Protein

Product Code. 15980303
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15980303

Brand: Invitrogen™ A30980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This MYLK4 recombinant human protein (full length) was expressed in insect cells. MYLK4 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for MYLK4 is NP_001012418.2 .

MYLK4 serine/threonine kinase or myosin light chain kinase family member 4 is a member of the CAMK kinase family. It is one of four myosin light chain kinases noted in a comprehensive analysis of protein kinases encoded by the human genome. MLCKs are activated by Ca2+-calmodulin complexes and require Mg2+ or Ca2+ as a co-factor. It has 2 isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86YV6
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 150mM NaCl, 0.1mM EDTA, 0.1mM PMSF, 10mM glutathione, 25% glycerol, 0.25mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 340156
Molecular Weight (g/mol) 70.9 kDa
Name Human MYLK4, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias caMLCK like; MYLK4; Myosin light chain kinase family member 4; myosin light chain kinase family, member 4; SgK085; Sugen kinase 85
Common Name MYLK4
Gene Symbol MYLK4
KinaseFamily CAMK Unique Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.