missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYLK4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95118-0.02ml
This item is not returnable.
View return policy
Description
MYLK4 Polyclonal antibody specifically detects MYLK4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MYLK4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| caMLCK like, EC 2.7.11, EC 2.7.11.1, myosin light chain kinase family, member 4, SGK085, SgK085myosin light chain kinase family member 4, Sugen kinase 85 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 119-388 of human MYLK4 (NP_001012418.2) QVHKCEETATGLKLAAKIIKTRGMKDKEEVKNEISVMNQLDHANLIQLYDAFESKNDIVLVMEYVDGGELFDRIIDESYNLTELDTILFMKQICEGIRHMHQMYILHLDLKPENILCVNRDAKQIKIIDFGLARRYKPREKLKVNFGTPEFLAPEVVNYDFVSFPTDMWSVGVIAYMLLSGLSPFLGDNDAETLNNILACRWDLEDEEFQDISEEAKEFISKLLIKEKSWRISASEALKHPWLSDHKLHSRLNAQKKKNRGSDAQDFVTK | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 340156 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido