missing translation for 'onlineSavingsMsg'
Learn More

MYLK3 Antibody [HRP], Novus Biologicals™

Product Code. 30764003 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30764003 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30764003 Supplier Novus Biologicals Supplier No. NBP346974H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYLK3 Polyclonal antibody specifically detects MYLK3 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYLK3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias caMLCK, Cardiac-MyBP-C-associated Ca/CaM kinase, EC 2.7.11, EC 2.7.11.18, MGC126319, MGC126320, MLC kinase, MLCK2, MLCKcardiac-MyBP-C associated Ca/CaM kinase, myosin light chain kinase 3, putative myosin light chain kinase 3
Host Species Rabbit
Immunogen myosin light chain kinase 3. (Uniprot# Q32MK0)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 91807
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.