missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYLK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115609
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MYLK2.
MYLK2 (skMLCK) is a calcium/calmodulin-dependent serine/threonine kinase expressed exclusively in adult skeletal muscle. MYLK2 encodes a myosin light chain kinase, a calcium/calmodulin dependent enzyme, that is exclusively expressed in adult skeletal muscle.
Specifications
| MYLK2 | |
| Polyclonal | |
| Unconjugated | |
| MYLK2 | |
| 9830004H17Rik; BB138278; Klmc; KMLC; MLCK; MLCK2; Mylk2; myosin light chain kinase 2; myosin light chain kinase 2, skeletal muscle; myosin light chain kinase 2, skeletal/cardiac muscle; myosin, light polypeptide kinase 2; myosin, light polypeptide kinase 2, skeletal muscle; skMLCK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 117558, 228785, 85366 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P20689, Q8VCR8, Q9H1R3 | |
| MYLK2 | |
| A synthesized peptide derived from human MYLK2(Accession Q9H1R3), corresponding to amino acid residues P264-K314. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto