missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYLK2 Monoclonal antibody specifically detects MYLK2 in Human, Mouse samples. It is validated for Western Blot, ELISA, KnockDown
Specifications
Specifications
| Antigen | MYLK2 |
| Applications | Western Blot, ELISA, KnockDown |
| Classification | Monoclonal |
| Clone | 2G1 |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_149109 |
| Gene Alias | EC 2.7.11, EC 2.7.11.18, KMLC, MLCK2MLCK, myosin light chain kinase 2, myosin light chain kinase 2, skeletal/cardiac muscle, skeletal muscle, skMLCK |
| Host Species | Mouse |
| Immunogen | MYLK2 (NP_149109, 161 a.a. ∽ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KLLAKKPPSEASELTFEGVPMTHSPTDPRPAKAEEGKNILAESQKEVGEKTPGQAGQAKMQGDTSRGIEFQAVPSEKSEVGQALCLTAREEDCFQILDDC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?