missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MYLK (MLCK), GST Tag Recombinant Protein

Product Code. 10328682
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10328682

Brand: Invitrogen™ PV4350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

MYLK (MLCK) is a serine/threonine kinase that phosphorylates myosin regulatory light chains to assist in the production of contractile activity (myosin-actin interaction). MLCK is responsible for smooth muscle contraction via phosphorylation of a specific serine in the N-terminus of myosin light chains (MLC), an event that facilitates myosin interaction with actin filaments. It is a central determinant in the development of vascular permeability and tissue edema formation. In the nervous system it has been shown to control the growth initiation of astrocytic processes in culture and to participate in transmitter release at synapses formed between cultured sympathetic ganglion cells. MLCK acts as a critical participant in signaling sequences that result in fibroblast apoptosis. Smooth muscle and non-muscle isozymes are expressed in a wide variety of adult and fetal tissues and in cultured endothelium with qualitative expression appearing to be neither tissue- nor development-specific. Non-muscle isoform 2 is the dominant splice variant expressed in various tissues. The Telokin isoform, which binds calmodulin, has been found in a wide variety of adult and fetal tissues. MLCK is probably down-regulated by phosphorylation. The protein contains 1 fibronectin type III domain and 9 immunoglobulin-like C2-type domains.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15746
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 50% glycerol, 2mM DTT, 0.02% Triton X-100, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 4638
Molecular Weight (g/mol) 67.3 kDa
Name Human MYLK (MLCK), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 155 kda myosin light chain kinase homolog; 9530072E15Rik; A clone isolated from a chicken gizzard cDNA library which has high homology with the 3'-end sequence of the chicken gizzard myosin light chain kinase (MLCK) cDNA; The 5'-leader seqeuence derives from the intronic sequence of the MLCK gene.; A930019C19Rik; AAT7; AW489456; kinase related protein; kinase-related protein; KRP; KRP/Telokin; MLCK; MLCK1; MLCK108; MLCK210; MSTP083; mylk; MYLK1; myosin light chain kinase; myosin light chain kinase, smooth muscle; Myosin light chain kinase, smooth muscle, deglutamylated form; myosin, light polypeptide kinase; nmMlck; smMLCK; smooth muscle myosin light chain kinase; smooth muscle myosin light chain kinase (61-kDa active fragment); smooth muscle myosin light chain kinase precursor (EC 2.7.2.37); telokin; unnamed protein product
Common Name MYLK
Gene Symbol MYLK
KinaseFamily MLCK Kinase Family
Protein Length 1428-1771
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1428-1771
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser/Thr Kinases: CAMK Group
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Catalytic
Form Liquid
Research Category Drug Development
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.