missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYLIP Polyclonal antibody specifically detects MYLIP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | MYLIP |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | band 4.1 superfamily member BZF1, BZF1, cellular modulator of immune recognition (c-MIR), EC 6.3.2.-, Idol, IDOLmyosin regulatory light chain-interacting protein, Inducible degrader of the LDL-receptor, MIRE3 ubiquitin-protein ligase MYLIP, myosin regulatory light chain interacting proteinE3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor |
| Host Species | Rabbit |
| Immunogen | myosin regulatory light chain interacting protein. (Uniprot# Q8WY64) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?