Learn More
Invitrogen™ MYL9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 720325
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey and Cat.
Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.
Specifications
| MYL9 | |
| Polyclonal | |
| Unconjugated | |
| MYL9 | |
| 20 kDa myosin light chain; AI327049; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; MGC3505; MLC2; MLC-2; MLC20; MLC-2C; MRLC1; myl9; myl9.L; myl9b; myl9l; Mylc2c; myosin light chain 2; myosin light chain 9; myosin light chain 9 L homeolog; myosin light chain, regulatory C; myosin regulatory light chain; myosin regulatory light chain 1; myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; myosin regulatory light polypeptide 9; myosin regulatory light polypeptide 9b; myosin RLC; myosin, light chain 9, regulatory; myosin, light chain 9b, regulatory; myosin, light polypeptide 9, regulatory; Myrl2; RLC-C; smooth muscle major isoform; unnamed protein product; XELAEV_18003093mg; zgc:77916 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10398, 98932 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4 | |
| P24844, Q9CQ19 | |
| MYL9 | |
| Peptides corresponding to Human MYL9 (aa 3-16, 111-124). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.