missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL9 [p Thr18, p Ser19] Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33583-100ul
This item is not returnable.
View return policy
Description
MYL9 Monoclonal antibody specifically detects MYL9 in Human samples. It is validated for ELISA,Western Blot
Specifications
| MYL9 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:7000, ELISA Recommended starting concentration is 1 μg/mL | |
| LC2020 kDa myosin light chain, MLC2Myosin RLC, MRLC1MLC-2C, myosin regulatory light chain 1, Myosin regulatory light chain 2, smooth muscle isoform, Myosin regulatory light chain 9, Myosin regulatory light chain MRLC1, myosin regulatory light polypeptide 9, myosin, light chain 9, regulatory, myosin, light polypeptide 9, regulatory, MYRL2MGC3505 | |
| A synthetic phosphorylated peptide around T18/S19 of human MYL9 (NP_006088.2).,, Sequence:, RATSNV | |
| 100 μL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 10398 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction