missing translation for 'onlineSavingsMsg'
Learn More

MYL7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16857154 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16857154 150 μL 150µL
16867154 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16857154 Supplier Proteintech Supplier No. 172831AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Encoded by the MYL7 gene, myosin regulatory light chain 7, also designated Myosin regulatory light chain 2, atrial isoform (MLC-2a), is part of a hexameric complex of two heavy chains and four light chains predomitly expressed in adult atrial muscle. Myosin regulatory light chain 7 binds calcium and has been shown to be a useful molecular marker for cardiac chamber specification. The co-expression of myosin regulatory light chain 7 and myosin light chain 2 (MLC2v) in the outflow tract and atrioventricular canal, together with the single expression in the atrial (MYL7) and ventricular (MYL2) myo-cardium, permits the delineation of their boundaries. At the amino acid level there is 95% homology between the human and mouse myosin regulatory light chain 7 sequences.

Specifications

Antigen MYL7
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYL7
Gene Accession No. Q01449, Q9QVP4
Gene Alias MLC 2a, MLC2a, MYL2A, MYL7, MYLC2A, Myosin light chain 2a
Gene Symbols Myl7
Host Species Rabbit
Immunogen MYL7 Fusion Protein Ag11058
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17898, 289759, 58498
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.