missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL6B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558579
This item is not returnable.
View return policy
Description
Immunogen sequence: APPAKTKAEP AVPQAPQKTQ EPPVDLSKVV IEFNKDQLEE FKE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in both slow-twitch skeletal muscle and in nonmuscle tissue.
Specifications
| MYL6B | |
| Polyclonal | |
| Unconjugated | |
| Myl6b | |
| 5730437E04Rik; BC037527; MLC1SA; MYL6B; myosin alkali light chain 1 slow a; myosin light chain 1 slow a; myosin light chain 1 slow-twitch muscle A isoform; myosin light chain 1, slow-twitch muscle A isoform; myosin light chain 6B; myosin, light chain 6B, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6B; myosin, light polypeptide 6B, alkali, smooth muscle and non-muscle; sm; smooth muscle and non-muscle myosin alkali light chain 6B; smooth muscle and nonmuscle myosin light chain alkali 6B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P14649 | |
| Myl6b | |
| Recombinant protein corresponding to Human MYL6B. Recombinant protein control fragment (Product #RP-97888). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction