missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106803
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MYL6.
Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain that is expressed in smooth muscle and non-muscle tissues. Genomic sequences representing several pseudogenes have been described and two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq, Jul 2008].
Specifications
| MYL6 | |
| Polyclonal | |
| Unconjugated | |
| MYL6 | |
| 17 kDa myosin light chain; ESMLC; LC17; LC17A; LC17B; LC17-GI; LC17-NM; MLC1SM; MLC-3; MLC3NM; MLC3SM; Myl6; Myln; myosin light chain 3; myosin light chain 6; myosin light chain A3; Myosin light chain alkali 3; myosin light chain, alkali, nonmuscle; myosin light polypeptide 6; myosin, light chain 6, alkali, smooth muscle and non-muscle; myosin, light polypeptide 6, alkali, smooth muscle and non-muscle; smooth muscle and nonmuscle myosin light chain alkali 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17904, 4637, 685867 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P60660, Q60605, Q64119 | |
| MYL6 | |
| A synthesized peptide derived from human MYL6(Accession P60660), corresponding to amino acid residues E59-I109. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction