missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | MYL6 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MYL6 Polyclonal antibody specifically detects MYL6 in Human, Mouse samples. It is validated for Western BlotSpecifications
| MYL6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS (pH 7.3), 50% glycerol | |
| 4637 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ESMLC, LC17, LC17A, LC17B, LC17-GI, LC17-NM, MLC1SM, MLC-3, MLC3NM, MLC3SM, Myosin light chain 3, Myosin light chain A3, Myosin light chain alkali 3, myosin light polypeptide 6,17 kDa myosin light chain, myosin, light chain 6, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6, alkali, smooth muscle and non-muscle, Smooth muscle and nonmuscle myosin light chain alkali 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-151 of human MYL6 (NP_066299.2). MCDFTEDQTAEFKEAFQLFDRTGDGKILYSQCGDVMRALGQNPTNAEVLKVLGNPKSDEMNVKVLDFEHFLPMLQTVAKNKDQGTYEDYVEGLRVFDKEGNGTVMGAEIRHVLVTLGEKMTEEEVEMLVAGHEDSNGCINYEAFVRHILSG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title