missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL5 Monoclonal Antibody (3D12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524578
This item is not returnable.
View return policy
Description
Peptide Sequence: RKTKKKEGGA LRAQRASSNV FSNFEQTQIQ EFKEAFTLMD QNRDGFIDKE DLKDTYASLG KTNVKDDELD AMLKEASGPI NFTMFLNLFG EKL.
This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia.
Specifications
| MYL5 | |
| Monoclonal | |
| Unconjugated | |
| MYL5 | |
| MYL5; MYLC2; myLC-2; Myosin light chain 5; myosin regulatory light chain 5; myosin, light chain 5, regulatory; myosin, light polypeptide 5, regulatory; superfast myosin regulatory light chain 2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4636 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3D12 | |
| PBS with no preservative; pH 7.4 | |
| Q02045 | |
| MYL5 | |
| MYL5 (NP_002468, 4 a.a. ~ 96 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction