missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL4 Monoclonal Antibody (OTI5A2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526892
This item is not returnable.
View return policy
Description
MYL4 Monoclonal Antibody for Western Blot, IHC (P)
Regulatory light chain of myosin. Does not bind calcium.
Specifications
| MYL4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P12829 | |
| MYL4 | |
| Human recombinant protein fragment corresponding to amino acids 2-197 of MYL4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5A2 | |
| Unconjugated | |
| MYL4 | |
| ALC1; AMLC; ELC; ELC1a; GT1; MLC1; MLC1a; MLC1EMB; MYL4; Myla; myosin light chain 1, atrial isoform; myosin light chain 1, atrial/fetal isoform; myosin light chain 1, embryonic muscle/atrial isoform; Myosin light chain 4; myosin light chain alkali GT-1 isoform; myosin light chain, alkali, cardiac atria; myosin, atrial/fetal muscle, light chain; myosin, light chain 4; myosin, light chain 4, alkali; atrial, embryonic; myosin, light polypeptide 4; myosin, light polypeptide 4, alkali; atrial, embryonic; PRO1957 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4635 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction