missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-46978
This item is not returnable.
View return policy
Description
MYL4 Polyclonal antibody specifically detects MYL4 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| MYL4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:20-1:200 | |
| ALC1, AMLC, GT1, MLC1, Myosin light chain 1, embryonic muscle/atrial isoform, myosin light chain 4, Myosin light chain alkali GT-1 isoform, myosin, atrial/fetal muscle, light chain, myosin, light chain 4, alkali; atrial, embryonic, myosin, light polypeptide 4, alkali; atrial, embryonic, PRO1957 | |
| myosin, light chain 4, alkali; atrial, embryonic. (Uniprot# P12829) | |
| 100 μg | |
| Vision | |
| 4635 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| PBS, 50% Glycerol, pH (7.3) | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction