missing translation for 'onlineSavingsMsg'
Learn More

MYL4 Antibody (1A11-C8), Novus Biologicals™

Product Code. 18372888 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372888 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372888 Supplier Novus Biologicals Supplier No. H00004635M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MYL4 Monoclonal antibody specifically detects MYL4 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYL4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A11-C8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH30228
Gene Alias ALC1, AMLC, GT1, MLC1, Myosin light chain 1, embryonic muscle/atrial isoform, myosin light chain 4, Myosin light chain alkali GT-1 isoform, myosin, atrial/fetal muscle, light chain, myosin, light chain 4, alkali, atrial, embryonic, myosin, light polypeptide 4, alkali, atrial, embryonic, PRO1957
Host Species Mouse
Immunogen MYL4 (AAH30228, 1 a.a. ~ 197 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAPKKPEPKKEAAKPAPAPAPAPAPAPAPAPEAPKEPAFDPKSVKIDFTADQIEEFKEAFSLFDRTPTGEMKITYGQCGDVLRALGQNPTNAEVLRVLGKPKPEEMNVKMLDFETFLPILQHISRNKEQGTYEDFVEGLRVFDKESNGTVMGAELRHVLATLGEKMTEAEVEQLLAGQEDANGCINYEAFVKHIMSG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4635
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.