missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582553
This item is not returnable.
View return policy
Description
Immunogen sequence: KPEPKKDDAK AAPKAAPAPA PPPEPERPKE VEFDASKIKI EFTPEQIEEF KEAFMLFDRT PKCEM Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 81%.
MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.
Specifications
| MYL3 | |
| Polyclonal | |
| Unconjugated | |
| MYL3 | |
| Cardiac myosin light chain 1; CMH8; CMLC1; MLC1s; MLC1SB; Mlc1v; MLClV; MLC-lV/sb; MYL3; Mylc; Mylc1v; Myosin alkali light chain 1, ventricular; Myosin alkali light chain 1, ventricular/slow skeletal muscle isoform; Myosin light chain 1, slow-twitch muscle B/ventricular isoform; myosin light chain 3; Myosin light chain 3, alkali, cardiac ventricles; myosin light chain, alkali, cardiac ventricles; myosin, light chain 3, alkali; ventricular, skeletal, slow; myosin, light polypeptide 3; myosin, light polypeptide 3, alkali; ventricular, skeletal, slow; rVMLC1; slow skeletal; slow skeletal ventricular myosin alkali light chain 3; ventricular; ventricular myosin alkali light chain; Ventricular myosin light chain 1; ventricular, skeletal, slow; ventricular/slow twitch myosin alkali light chain; VLC1; VLCl | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4634 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P08590 | |
| MYL3 | |
| Recombinant protein corresponding to Human MYL3. Recombinant protein control fragment (Product #RP-91305). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction